Would you like to log in again?
8095 Matrix Spike Mix B (7 Components), Restek
Supplier: Restek
31610
75835-706EA 42.68 USD InStock
75835-706
8095 Matrix Spike Mix B (7 Components), Restek
Standards Organic Standards
The mix consists of, 3,5-dinitroaniline, nitrobenzene, nitroglycerin, 2-nitrotoluene, 3-nitrotoluene, 4-nitrotoluene, PETN.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty