Would you like to log in again?
Chloroprene, Restek
Supplier: Restek
30238
75835-550EA 43.65 USD InStock
75835-550
Chloroprene, Restek
Standards Organic Standards
A pure, quantitative chloroprene solution, specially stabilised in purge-and-trap grade methanol to allow dilutions for working standards in unmodified purge-and-trap methanol. the entire preparation procedure is performed under carefully monitored conditions to prevent any contamination or polymerisation of the highly reactive, neat chloroprene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty