Would you like to log in again?
8270/Appendix IX Kit, Restek
Supplier: Restek
31815
75834-776KT 311.37 USD InStock
75834-776
8270/Appendix IX Kit, Restek
Standards Organic Standards
This kit contains 1 ml each of these mixtures: 8270 MegaMix standard, 605 benzidines calibration mix, appendix IX mix #1, revised (methapyrilene is not included in this revised standard), methapyrilene standard, appendix IX Mix #2.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Ordering information: Quantity discounts not available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty