Would you like to log in again?
Formaldehyde-DNPH Mix, Restek
Supplier: Restek
31837
75834-810EA 71.78 USD InStock
75834-810
Formaldehyde-DNPH Mix, Restek
Standards Organic Standards
This mix with acetonitrile solvent suitable for HPLC analysis.
  • High concentration
Sampling for airborne carbonyl compounds in EPA Method TO-11A and other methods is based on a reaction between the target compounds and 2,4-dinitrophenylhydrazine (DNPH), coated on a silica gel adsorbent. Meet the needs of investigators analyzing only for formaldehyde.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty