Would you like to log in again?
Retention Time Marker, Restek
Supplier: Restek
30483
75834-396EA 40.74 USD InStock
75834-396
Retention Time Marker, Restek
Standards Organic Standards
Used during initial sample screening to determine retention time windows for each petroleum product
Gasoline generally elutes in the window from C6 to C10 (or C12), and diesel fuel from C10 (or C12) to C24 (or C28). Retention above C24 (or C28) indicates oil or lubricant contamination.
3 components
Certifications: Retention Time Standards
Ordering information: State of Alaska
Method and regulatory information is available from:
Alaska Department of Environmental Conservation
410 Willoughby Avenue
Juneau, AK 99801-1795
Phone: (907)465-5203
Fax: (907)465-5218
www.state.ak.us/dec/dspar/stp_home.htm
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty