Would you like to log in again?
CCME PHC Calibration Mix (3 Components), Restek
Supplier: Restek
31684
75834-738EA 43.65 USD InStock
75834-738
CCME PHC Calibration Mix (3 Components), Restek
Standards Organic Standards
This calibration mix consist of three components: (C10) n-Decane, (C16) n-Hexadecane and (C34) n-Tetratriacontane.
  • Meets CCME 2001 Petroleum Hydrocarbons in Soil Method—Tier 1
  • Primary reference calibration standards for quantification of four fractions.
Shelf life 6 to 36 months.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty