Would you like to log in again?
Quality Control Standard Mixture (2 Components), Restek
Supplier: Restek
31631
75835-730EA 41.71 USD InStock
75835-730
Quality Control Standard Mixture (2 Components), Restek
Standards Organic Standards
For GC analysis of total petroleum hydrocarbons (TPH) in water.
  • Environmentally safer than previous methods
  • Calibration standard available as diesel #2/motor oil and diesel #2/mineral oil
  • Total hydrocarbon concentration is 1000 μg/ml
The mix consists of diesel #2 (additive-free), mineral oil (additive-free [ USP grade] bp 391 - 522 or C24 - C40 retention time range).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty