Would you like to log in again?
System Performance Test Standard Mixture of n-Alkanes (16 Components), Restek
Supplier: Restek
31678
75834-728EA 89.24 USD InStock
75834-728
System Performance Test Standard Mixture of n-Alkanes (16 Components), Restek
Standards Organic Standards
This standard mixture cobtains: (C10) n-Decane, (C12) n-Dodecane, (C14) n-Tetradecane, (C16) n-Hexadecane, (C18) n-Octadecane, (C20) n-Eicosane, (C22) n-Docosane, (C24) n-Tetracosane, (C26) n-Hexacosane, (C28) n-Octacosane, (C30) n-Triacontane, (C32) n-Dotriacontane, (C34) n-Tetratriacontane, (C36) n-Hexatriacontane, (C38) n-Octatriacontane and (C40) n-Tetracontane.
Shelf life 6 to 96 months.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty