Would you like to log in again?
Extraction Solvent Stock Solution #1 (2 Components), Restek
Supplier: Restek
31679
75834-730EA 45.59 USD InStock
75834-730
Extraction Solvent Stock Solution #1 (2 Components), Restek
Standards Organic Standards
This extraction solvent stock solution contains: n-decane (C10), n-tetracontane (C40).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty