Would you like to log in again?
Extraction Solvent Stock Solution #2 (2 Components), Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
31680
75834-732EA 71.78 USD InStock
75834-732
Extraction Solvent Stock Solution #2 (2 Components), Restek
Standards Organic Standards
This extraction solvent stock solution contains: n-decane (C10) (20 µl/l), n-tetracontane (C40) (20 mg/l).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty