Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
USP <467> Calibration Mix #7, Restek
Supplier: Restek
36009
75835-430EA 42.68 USD InStock
75835-430
USP <467> Calibration Mix #7, Restek
Standards Organic Standards
The Class 1 mixtures below reflect the requirements of USP23/NF18 effective January 1, 1995 to December 31, 1999
While these mixtures do not meet the current USP guidelines, many still use these mixtures to obtain a detectable benzene peak for the direct injection methods, Method I and Method V.
4 components
Certifications: USP <467>
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty