Would you like to log in again?
USP <467> Calibration Mixture #3 (5 components), Restek
Supplier: Restek
36004
75835-424EA 40.74 USD InStock
75835-424
USP <467> Calibration Mixture #3 (5 components), Restek
Standards Organic Standards
The mixture consists of benzene (100 μg/ml), chloroform 50 μg/ml, 1,4-dioxane (100 μg/ml), methylene chloride (dichloromethane) (500 μg/ml) and trichloroethene (100 μg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty