Would you like to log in again?
Standard Mixture Stock Solution (2 Components), Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
31630
75835-728EA 42.68 USD InStock
75835-728
Standard Mixture Stock Solution (2 Components), Restek
Standards Organic Standards
For GC analysis of total petroleum hydrocarbons (TPH) in water.
  • Calibration standard available as diesel #2/motor oil and diesel #2/mineral oil
The mix consists of diesel #2 (additive-free) and mineral oil (additive-free [ USP grade] bp 325 - 460 or C18 - C32 retention time range).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty