Would you like to log in again?
Oxygenates Standard, Restek
Supplier: Restek
30619
75834-512EA 42.68 USD InStock
75834-512
Oxygenates Standard, Restek
Standards Organic Standards
This standard consists of five components.
This standard consists of five components.
Components: tert-Amyl ethyl ether (2000 µg/ml), tert-Amyl methyl ether (2000 µg/ml), tert-Butanol (10000 µg/ml), Diisopropyl ether (2000 µg/ml) and Ethyl-tert-butyl ether (2000 µg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty