Would you like to log in again?
European Pharmacopoeia/ICH Q3C(M) Class 2 Mix A, Revised, Restek
Supplier: Restek
36274
75835-460EA 50.44 USD InStock
75835-460
European Pharmacopoeia/ICH Q3C(M) Class 2 Mix A, Revised, Restek
Standards Organic Standards
The analysis of residual solvents in pharmaceutical products has changed, particularly for products being sold into Europe
The International Conference on Harmonization (ICH) Guidelines for Residual Solvents is becoming the international standard and is being adopted by more pharmacopoeias, including the United States Pharmacopeia, every year. The ICH method and compound list is more extensive than any method previously used and poses new challenges. Compounds in Class 1 are solvents considered to be of highest risk and to be avoided in pharmaceutical manufacturing. Use of Class 2 compounds is to be limited, as they pose a lower, but present, threat to health. Compounds in Class 3 pose the lowest toxic potential and may be used routinely in manufacturing.
14 components
Certifications: European Pharmacopoeia Method
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty