Would you like to log in again?
Free Fatty Acids Test Standard (6 Components), Restek
Supplier: Restek
35272
75835-390EA 69.84 USD InStock
75835-390
Free Fatty Acids Test Standard (6 Components), Restek
Standards Organic Standards
The free fatty acids test standard consists of six components they are (C4:0) butyric acid, acetic acid, isobutyric acid, isovaleric acid, propionic acid and valeric acid.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty