Would you like to log in again?
NJDEP EPH 10/08 Rev. 2 Aliphatics Calibration Standard (20 Components), Restek
Supplier: Restek
30540
75834-476EA 61.11 USD InStock
75834-476
NJDEP EPH 10/08 Rev. 2 Aliphatics Calibration Standard (20 Components), Restek
Standards Organic Standards
Consists of 20 components, those are n-nonane, n-decane, n-dodecane, n-tetradecane, n-hexadecane, n-octadecane, n-eicosane, n-heneicosane, n-docosane, n-tetracosane, n-hexacosane, n-octacosane, n-triacontane, n-dotriacontane, n-tetratriacontane, n-hexatriacontane, n-octatriacontane, n-tetracontane, 2-methylnaphthalene and naphthalene.
20 components
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty