Would you like to log in again?
Pharmaceuticals Mix #1 (8 Components), Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
31116
75834-620EA 64.99 USD InStock
75834-620
Pharmaceuticals Mix #1 (8 Components), Restek
Standards Organic Standards
This pharmaceuticals mix #1 contain eight components: Acetaminophen, Caffeine, Carbamazepine, Ciprofloxacin HCL, Erythromycin USP, Fluoxetine HCl, Sulphamethoxazole and Trimethoprim.
Shelf life 18 months.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty