Would you like to log in again?
DHA PONA Standard, Restek
Supplier: Restek
30731
75834-590EA 1117.43 USD InStock
75834-590
DHA PONA Standard, Restek
Standards Organic Standards
ASTM Methods D6729, D6730, and D6733 (Determination of Individual Components in Spark Ignition Engine Fuels) are designed for the determination of the individual hydrocarbons present in spark ignition engine fuels, as well as fuel blends containing oxygenates such as methyl tert-butyl ether, ethyl tert-butyl ether, tert-butanol, ethanol, etc.
This standard is a qualitative mixture of various gasoline and refinery materials prepared to provide nearly every component that may be encountered in feedstock and finished gasolines. Some oxygenates have been added to allow this blend to be used by refineries for detailed hydrocarbon analysis (DHA) method setup and to fulfill requirements such as ASTM Methods D6729, D6730, and D6733.
188 components
Certifications: ASTM Methods D6729, D6730, and D6733 (Determination of Individual Components in Spark Ignition Engine Fuels)
Ordering information: Quantity discounts not available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty