Would you like to log in again?
EU 15+1 PAH Standard, Restek
Supplier: Restek
32470
75835-040EA 219.22 USD InStock
75835-040
EU 15+1 PAH Standard, Restek
Standards Organic Standards
  • Formulated to contain the 16 major European Food Safety Authority (EFSA) PAH compounds
  • High concentration (100 μg/mL) solution provides value and flexibility. Dilute as needed for fortification and calibration mixes
  • Low-volatility toluene solvent provides longer shelf life, which helps ensure more accurate results
  • Broad solvent miscibility—compatible with acetonitrile, the QuEChERS solvent
  • Certified reference material (CRM) meets strict ISO quality requirements
  • Pair with Rxi-PAH GC column for easy separation of critical PAH compounds from common interferences in food and environmental samples: www.restek.com/rxi-pah
  • HPLC compatible when diluted in appropriate solvent and analyzed with Pinnacle II PAH columns or UHPLC compatible on the Pinnacle DB PAH column.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty