Would you like to log in again?
Pharmaceutical Impurities Standard, Restek
Supplier: Restek
30732
75834-592EA 60.14 USD InStock
75834-592
Pharmaceutical Impurities Standard, Restek
Standards Organic Standards
  • Features two common alkyl halides—chloromethane and chloroethane—that can form during drug synthesis
  • High-concentration (5,000 μg/mL) solution maximizes flexibility; ideal for analyzing pharmaceutical genotoxic impurities (GTIs)
  • Quantitatively tested to confirm composition and stability; detailed support documentation provided
  • Certified reference material (CRM) manufactured and QC-tested in Restek’s ISO-accredited labs satisfies your ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty