Would you like to log in again?
BioClean™ Maxima™ Sterile Latex Gloves, Ansell
Supplier: Nitritex
BioClean Maxima™ BioClean™
BLLS60 BLLS75 BLLS90 BLLS85 BLLS100 BLLS60 BLLS70 BLLS80 BLLS75 BLLS65 BLLS70 BLLS65 BLLS80 BLLS85 BLLS90 BLLS100
75834-024CS 2315.34 USD InStock
CA75834-022 75834-024 CA75834-100 75834-102 CA75834-024 75834-016 CA75834-102 75834-018 CA75834-016 CA75834-018 75834-020 75834-098 75834-022 CA75834-020 75834-100 CA75834-098
BioClean™ Maxima™ Sterile Latex Gloves, Ansell
Gloves
BioClean™ Maxima™ Sterile Latex Gloves offer full arm protection.
  • Exceptional flexibility and comfort
  • Shoulder length
  • Textured for improved grip
  • Double-donnable
  • Non-particulating EasyTear™ packaging
BioClean™ Maxima™ Sterile Latex Glove is specifically manufactured to provide shoulder length protection without sacrificing the exceptional flexibility and comfort provided by latex. This natural-coloured, hand-specific, anatomically shaped glove features a textured surface for enhanced grip and a beaded cuff for stability on the arm.
Certifications: Manufactured in a facility holding ISO 9001: 2008 and 89/686/EEC Article 11b certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Directive 89/686/EEC.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of Glove Standard EN 420:2003 and EN 374-1, 2 & 3:2003.
Physical properties comply with European Medical Glove Standard EN 455-2:2009.
Sterilized in accordance with ISO 11137-1:2006.
ISO Class 5 compatible.
Packaging: Gloves are packaged one pair per inner PE wallet; one wallet per sealed EasyTear™ PE pouch; 10 pouches per sealed outer PE bag; 10 outer bags per lined carton (100 pairs).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty