Would you like to log in again?
Accessories for E-A-Rsoft™ FX™ Earplugs, 3M™
Supplier: 3M
E-A-RSoft™
7100038299 7100088422
10016-972CS 1257.92 USD InStock
10016-972 10204-906
Accessories for E-A-Rsoft™ FX™ Earplugs, 3M™
Earplugs Disposable Earplugs
Each refill bottle/box easily mounts into a dispenser, making the system easy for workers to access earplugs for hearing protection.
  • E-A-Rsoft™ FX™ Earplugs use soft, slow recovery foam
  • Noise Reduction Rating (NRR) of 33 dB
  • Used in many industries
These earplugs are test compatible with the E-A-Rfit™ Validation System. Moisture resistance helps maintain a consistent expansion time, even when humidity is present. Flame resistance helps protect against potential arc flash situations and provides for safer storage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty