Would you like to log in again?
Accessories for E-A-Rsoft™ SuperFit™ Earplugs, 3M™
Supplier: 3M
E-A-RSoft™
7000042881
10016-978CS 908.11 USD InStock
10016-978
Accessories for E-A-Rsoft™ SuperFit™ Earplugs, 3M™
Earplugs Disposable Earplugs
E-A-Rsoft™ SuperFit™ foam earplugs seal the earcanal with soft, slow-recovery foam for excellent noise reduction and comfort.
  • Refill container designed for use with the One Touch™ dispenser
  • One gentle turn delivers uncorded foam earplugs
  • Regular size fits most earcanals
  • Noise Reduction Rating (NRR) of 33 dB; CSA Class AL
  • Test compatible with 3M™ E-A-Rfit™ Validation System
The soft, pliable foam on this uncorded foam earplug features an orange fitting ring that provides a guideline for proper fitting
The NRR may overestimate the hearing protection provided during typical use. 3M™ recommends reducing the NRR by 50% for estimating the amount of noise reduction provided.
Earplugs are easy to roll down and insert, then expand within the earcanal for a comfortable and secure fit.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty