Would you like to log in again?
Accessories for E-A-Rsoft™ Yellow Neon™ Blasts™ Earplugs, 3M™
Supplier: 3M
E-A-RSoft™
7000052750 7100038299
10204-904CS 405.05 USD InStock
10204-904 10016-964
Accessories for E-A-Rsoft™ Yellow Neon™ Blasts™ Earplugs, 3M™
Earplugs Disposable Earplugs
Yellow Neon™ Blasts™ earplugs seal the earcanal with soft, slow-recovery foam for excellent noise reduction and comfort.
  • Rapid release packaging
  • Regular size fits most ear canals
  • Noise Reduction Rating (NRR) of 33 dB; (ANSI S3.19-1974) CSA Class AL (CSA Z94.2)
  • Test compatible with 3M™ E-A-Rfit™ Validation System.
These uncorded foam earplugs feature soft, slow-recovery foam for comfort and outstanding noise reduction
The NRR may overestimate the hearing protection provided during typical use. 3M™ recommends reducing the NRR by 50% for estimating the amount of noise reduction provided.
Finished with exciting flame graphics, these soft foam earplugs are easy to roll down and insert. They expand within the earcanal for a comfortable and secure fit.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty