Would you like to log in again?
Accessories for E-A-R™ Classic™ Earplugs, 3M
Supplier: 3M
7100038299 7010384042 7100001857 7000052726
10204-936CS 411.98 USD InStock
10204-936 10016-974 12003-002 47744-957
Accessories for E-A-R™ Classic™ Earplugs, 3M
Earplugs Disposable Earplugs
These earplugs are stiff enough when compressed to make fitting easy, yet soft enough to be comfortable in the ear for extended wear.
  • Conforms to the shape of the earcanal to help create a comfortable yet snug noise barrier
  • Slow-recovery earplug foam is non-flammable and moisture resistant
  • ANSI S3.19-1974; CSA Class AL
  • Test compatible with 3M™ E-A-Rfit™ Validation System
Slow recovery, low pressure foam fits snugly to create a noise barrier that helps protect hearing from loud sounds
The E-A-R™ Classic™ Earplugs were the world’s first foam earplugs and are still one of the most widely used earplugs today.
Bright yellow color allows supervisors to conduct quick visual hearing protection compliance checks. Recommended applications include chipping, chiseling, drilling, furnace operations, grinding, heavy equipment operations, machining, pouring/casting, power fastening, riveting, sanding, sawing, and welding. These earplugs are ideal for use in agriculture, automotive, chemicals, construction, general manufacturing, healthcare, oil and gas, and pharmaceutical industries.
These regular size earplugs comfortably fit most people.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty