Would you like to log in again?
PELTOR™ Throat Microphone, 3M™ Company
Supplier: 3M
PELTOR™
7000127275
89006-120EA 877.59 USD InStock
89006-120
PELTOR™ Throat Microphone, 3M™ Company
Sound Meters
This noise-canceling throat microphone enables crisp, reliable communication in the 2-way communication headsets of the 3M™ PELTOR™ Series, specifically designed for industrial users in high-noise environments.
  • Reduces or eliminates noise in high-noise areas
  • Filters out soil and other variants for clear, reliable communication
  • Compatible with 3M™ PELTOR™ Series headsets
  • Works in conjunction with 2-way radio for crisp 2-way communication
  • Used with headsets specifically designed for high-noise environments
Get better communication with maximum attenuation with this noise-canceling microphone accompanying the 2-way radio communication headsets of the 3M™ PELTOR™ Series. The microphone has a soil-repelling filter which allows for clear, reliable communication.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty