Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ACC2 Inhibitor


AB142090-1MG 1717-5
75836-798EA 337.08 USD InStock
75836-798 75836-800
ACC2 Inhibitor
ACC2 Inhibitor
Cell-permeable, a potent and selective inhibitor of ACC2 (Acetyl-CoA Carboxylase) (IC₅₀ = 28 nM for hACC2 vs 210 nM for hACC1 in in vitro studies).


Formula: C₁₉H₂₁N₃O₃S₂
MW: 403.52 g/mol
UN: 3316
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty