Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Posaconazole

Supplier: BioVision
Synonyms: 4-(4-(4-(4-(((3R,5R)-5-((1H-1,2,4-Triazol-1-yl)methyl)-5-(2,4-difluorophenyl)tetrahydrofuran-3-yl)methoxy)phenyl)piperazin-1-yl)phenyl)-1-((2S,3S)-2-hydroxypentan-3-yl)-1H-1,2,4-triazol-5(4H)-one

AB285414-10MG AB285414-50MG
75837-092EA 245.13 USD InStock
75837-092 75837-094
Posaconazole
Posaconazole
Posaconazole is a broad-spectrum, second-generation triazole compound that displays antifungal activity.

Posaconazole inhibits the growth of fungi by inhibiting the lanosterol 14α-demethylase enzyme (IC₅₀s range from 7-200 nM), which results in a disruption the synthesis of ergosterol, a key sterol necessary for fungal cell membrane integrity and for the function of certain membrane-bound proteins...


Formula: C₃₇H₄₂F₂N₈O₄
MW: 700,79 g/mol
MDL Number: MFCD00941162
CAS Number: 171228-49-2
UN: 3316
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty