Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Rabbit IgG Antibody (AP (Alkaline Phosphatase))

Supplier: Promega
S3731
PAS3731EA 151.37 USD InStock
PAS3731
Anti-Rabbit IgG Antibody (AP (Alkaline Phosphatase))
Antibodies
Polyclonal secondary antibody raised in goat, immunoaffinity-purified using corresponding immobilized antigens and conjugated to alkaline phosphatase (AP) enzyme.

  • Secondary Antibody That Reacts with the Heavy Chains of Rabbit IgG
  • Extensively used and published in numerous research articles
  • Antibody is ready-to-use

Polyclonal secondary antibody raised in goat, immunoaffinity-purified using corresponding immobilized antigens and conjugated to alkaline phosphatase (AP) enzyme. The product (unless otherwise noted) is supplied as a 1mg/ml solution. The Anti-Rabbit IgG (Fc), AP Conjugate antibody reacts with the heavy chains of rabbit IgG but not with the light chains. As with all antibodies, in certain applications some species-dependent antigen-dependent cross-reactivity may be observed. A starting working dilution of 1:2,500 is suggested for most Western blot, dot blot and ELISA applications. The optimum concentration of secondary antibody depends on the application and will need to be empirically determined.

Type:
Antigen: IgG
Clonality:
Clone:
Conjugation: AP (Akaline Phosphatase)
Epitope:
Host:
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty