Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Human IgG Antibody (AP (Alkaline Phosphatase))

Supplier: Promega
S3821
PAS3821EA 148.88 USD InStock
PAS3821
Anti-Human IgG Antibody (AP (Alkaline Phosphatase))
Antibodies
Polyclonal secondary antibody raised in goat, immunoaffinity-purified using corresponding immobilized antigens and conjugated to alkaline phosphatase (AP) enzyme. The human antibodies react with all IgG subclasses.

  • Secondary Antibody That Binds to Both Heavy and Light Chains for All Human IgG Subclasses
  • Also reacts with light chains on other human immunoglobulins
  • Extensively used and published in numerous research articles
  • Antibody is ready-to-use

Polyclonal secondary antibody raised in goat, immunoaffinity-purified using corresponding immobilized antigens and conjugated to alkaline phosphatase (AP) enzyme. The product (unless otherwise noted) is supplied as a 1mg/ml solution. The Anti-Human IgG (H+L), AP Conjugate antibody reacts with heavy and light chains of all subclasses of human IgG as well as with light chains on other human immunoglobulins; it displays minimal cross-reactivity to horse or bovine serum proteins. As with all antibodies, in certain applications some species-dependent antigen-dependent cross-reactivity may be observed. A starting working dilution of 1:2,500 is suggested for most Western blot, dot blot and ELISA applications. The optimum concentration of secondary antibody depends on the application and will need to be empirically determined.

Type:
Antigen: IgG
Clonality:
Clone:
Conjugation: AP (Akaline Phosphatase)
Epitope:
Host:
Isotype:
Reactivity: Rabbit

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty