Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Luciferase Goat Polyclonal Antibody

Supplier: Promega
G7451
PAG7451EA 357.6 USD InStock
PAG7451
Anti-Luciferase Goat Polyclonal Antibody
Antibodies
Anti-Luciferase pAb is a goat polyclonal antibody designed for use in immunocytochemistry and Western blotting.

  • Antibody That Recognizes Photinus pyralis Firefly Luciferase
  • Goat polyclonal IgG generated using 61kDa recombinant luciferase from North American firefly (Photinus pyralis)
  • Specific for firefly luciferase (Photinus pyralis); antibody does not cross-react with sea pansy (Renilla reniformis) luciferase
  • Antibody provided at 1mg/ml in PBS containing 50microg/ml gentamicin

Anti-Luciferase pAb is a goat polyclonal antibody designed for use in immunocytochemistry and Western blot applications. Anti-Luciferase pAb can detect luciferase enzyme expression in situ.

Type:
Antigen: Luciferase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: IgG
Host: Goat
Isotype:
Reactivity: Photinus pyralis (Firefly)

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty