Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HaloTag® Mouse Monoclonal Antibody

Supplier: Promega
G9211
PAG9211EA 542.14 USD InStock
PAG9211
Anti-HaloTag® Mouse Monoclonal Antibody
Antibodies
Anti-HaloTag Monoclonal Antibody is a mouse monoclonal antibody raised against the HaloTag protein, which can be used to detect HaloTag fusion proteins by Western blotting.

  • Monoclonal Antibody Raised Against the HaloTag(R) Protein
  • Little to no cross-reactivity with other non-HaloTag(R) proteins
  • Can detect as low as 0.5-1ng of HaloTag(R) fusion protein by Western blot
  • Supplied at a concentration of 1mg/ml

Anti-HaloTag Monoclonal Antibody is a mouse monoclonal antibody raised against the HaloTag protein, which can be used to detect HaloTag fusion proteins by Western blotting. The HaloTag platform addresses the need for flexibility in functional protein analysis for cell imaging, protein purification and protein pull-down applications.

Type:
Antigen: HaloTag®
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty