Would you like to log in again?
Helix pomatia Native Beta-glucuronidase
Supplier: EMD Millipore
347420-1MU
EM347420-1MUEA 427.13 USD InStock
EM347420-1MU
Helix pomatia Native Beta-glucuronidase
Enzymes
Useful for hydrolyzing urinary steroid conjugates prior to steriod assays.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty