Would you like to log in again?
Weighing Funnels, Wilmad-LabGlass
Supplier: SP Industries
LG-8315-102 LG-8315-100
14202-532EA 34.59 USD InStock
14202-532 14209-748
Weighing Funnels, Wilmad-LabGlass
Boats Weighing Boats
Made of borosilicate glass, this funnel provides a means of direct transfer to volumetric flasks, beakers, etc. The base of the funnel is flat to provide stability on the balance pan.
The base of the funnel is flat to provide stability on the balance pan.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty