Would you like to log in again?
Enteropeptidase Substrate, Fluorogenic, BioVision
Supplier: BioVision
AB285395-1MG AB285395-5MG
75837-746EA 552.96 USD InStock
75837-746 75837-744
Enteropeptidase Substrate, Fluorogenic, BioVision
Stains Histology Stains
Enteropeptidase substrate is a useful fluorogenic substrate for determining the activity of the enzyme enteropeptidase/enterokinase.
  • Recognizes a highly specific amino acid sequence ‘DDDDK’
  • Cleaves after the lysine residue
This enzyme recognizes a highly specific amino acid sequence ‘DDDDK’ and cleaves after the lysine residue releasing the AFC moiety that can be quantified using a fluorometer.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty