Would you like to log in again?
DiscoveryPak™, Stem Cell Fate Regulator Set X, BioVision
Supplier: BioVision
AB286221-1KIT
75837-940EA 819.72 USD InStock
75837-940
DiscoveryPak™, Stem Cell Fate Regulator Set X, BioVision
Cells
A convenient set of two small molecules useful for increasing the cloning efficiency in individualized human pluripotent stem cells (hPSCs).
  • Increase cloning efficiency
  • Convenient set of two molecules
The two modulators are: 5 mg of Pioglitazone and 5 mg of Y-27632 HCl.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty