Would you like to log in again?
3M™ Double Coated Tape 415 Clear, 3M Company
Supplier: 3M
7000048399
62111-234EA 81.83 USD InStock
62111-234
3M™ Double Coated Tape 415 Clear, 3M Company
Tapes
Offers a balance of initial adhesion and good holding power.
  • Excellent UV resistance. Good adhesion to a wide variety of materials such as HSE high surface energy, LSE low surface energy and metals
  • Good temperature and humidity resistance
  • High performance splicing in printing applications. Good on films and foils
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty