Would you like to log in again?
Scotch® Super 33+™ Vinyl Electrical Tape, 3M™
Supplier: 3M
7100002398
89235-840CS 709.31 USD InStock
89235-840
Scotch® Super 33+™ Vinyl Electrical Tape, 3M™
Tapes
The combination of elastic backing and aggressive adhesive provides moisture-tight electrical and mechanical protection with minimum bulk.
  • General-purpose vinyl insulating tape
  • Excellent resistance to: abrasion, moisture, alkalies, acid, copper corrosion
  • Resistant to varying weather conditions
Scotch® Super 33+™ Vinyl Electrical Tape is an UL Listed and CSA Certified insulating tape.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty