Would you like to log in again?
Agar Slant, Tubes, Northeast Lab Services
Supplier: Northeast Lab Services
T0601
101374-492PK 86.35 USD InStock
101374-492
Agar Slant, Tubes, Northeast Lab Services
Media for Microbiology Dehydrated Media for Microbiology
Agar Slant has a secure, glass screw cap.
  • Screw Cap
  • Glass Construction
  • Nine Milliliter Fill Volume
  • Prepared Medium
  • Sixteen by 125 mm Tube
This slant is contained in a 16 x 125 mm glass tube.
From simple fermentation tubes to highly specialized clinical diagnostic media, NEL produces media tubes for a wide variety of clients including private laboratories, food processors, clinics and hospitals, academic institutions, and government agencies.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty