Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sim Medium, Tubes, Northeast Lab Services

Supplier: Northeast Lab Services
T1720
101374-798PK 72 USD InStock
101374-798
Sim Medium, Tubes, Northeast Lab Services
Media for Microbiology Dehydrated Media for Microbiology
Sim Medium features a fill volume of eight milters and 16 x 125 mm tubes.

  • Prepared Medium
  • Glass Construction
  • Screw Cap
  • Fill Volume: 8 mL
  • Size: 16 x 125 mm

Other features include a screw cap and glass construction for easy visibility.

NEL produces media tubes for private laboratories, food processors, clinics and hospitals, academic institutions, government agencies, and more.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty