Would you like to log in again?
Cell Biology Hoagland's Modified Basal Salt Mixture, MP Biomedicals
Supplier: MP Biomedicals
092621820 092621822
IC2621822EA 29.02 USD InStock
IC2621822 IC2621820
Cell Biology Hoagland's Modified Basal Salt Mixture, MP Biomedicals
Media for Microbiology Dehydrated Media for Microbiology
Supports and facilitates plant growth.
  • Includes macro- and micronutrients
  • Plant Media
  • Soluble 1X in water
Formulation: Ammonium Phosphate 115.03 mg/L, Boric Acid 2.86 mg/L, Calcium Nitrate 656.4 mg/L, Cupric Sulfate5H2O 0.08 mg/L, Na 2 -EDTA 33.534 mg/L, Ferrous Sulfate7H2O 25.02 mg/L, Magnesium Sulfate 240.76 mg/L, Manganese Chloride· 4H2O 1.81 mg/L, Molybdenum Trioxide 0.016 mg/L, Potassium Nitrate 606.6 mg/L, Zinc Sulfate7H2O 0.22 mg/L.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty