Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cell Biology Murashige and Skoog Modified Vitamin Solution (1000X), MP Biomedicals

Supplier: MP Biomedicals
092625149
IC2625149EA 65.95 USD InStock
IC2625149
Cell Biology Murashige and Skoog Modified Vitamin Solution (1000X), MP Biomedicals
Media for Microbiology Dehydrated Media for Microbiology
Contains the vitamins as described by Murashige and Skoog (1962) modified with additional Thiamine.

  • Plant Media
  • Typical Working Concentration: 1 mL/ L of Medium
  • Soluble in water

Formula for solution (mg/L): Glycine (Free Base): 2000, myo-Inositol: 100,000, Nicotinic Acid (Free Acid): 500, PyridoxineHCI: 500, ThiamineHCI: 1000

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty