Would you like to log in again?
Portable Light Weight Stroboscopes with Rechargeable Battery, Nidec Shimpo
Supplier: Nidec-Shimpo
DT-725KIT-230V DT-725KIT
75836-600EA 1068.34 USD InStock
75836-600 75836-602
Portable Light Weight Stroboscopes with Rechargeable Battery, Nidec Shimpo
Tachometers
The DT-725 possess many of the same features as the famous DT-300 Series Stroboscopes, but are lighter in weight.
  • Exceptionally light weight design improves portability
  • External trigger allows unit to be automatically synchronized with equipment
  • Continuous duty cycle eliminates need to shut down for cooling
  • Easy adjust flash rate controller dial
  • Synchronous output enables strobe to drive other strobes
The DT-725 possess many of the same features as the famous DT-300 Series Stroboscopes, but are lighter in weight. Constructed of strengthened high impact plastic, these rugged units are exceptionally reliable and withstand the same harsh conditions as more expensive units. Employing its unique phase shifting feature, the DT-725 “freezes” rotating or reciprocating machinery for visual inspection of covered gear teeth, shafts, oil/fuel/ink injector patterns, and print media; all without stopping production.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty