Would you like to log in again?
Justrite® Blue Corrosive Wood Laminate Cabinet
Supplier: Justrite
24150
470113-032EA 4565.75 USD InStock
470113-032
Justrite® Blue Corrosive Wood Laminate Cabinet
Furniture Storage Cabinets
Durable and Highly Chemical-Resistant
  • Feature Haz-Alert™ reflective warning labels
  • Adjustable shelves
  • Adjustable feet for leveling
  • Double key set for safety and security
Safely store highly corrosive liquids such as phenol, nitric acid and sulfuric acid in these wood laminate storage cabinets. Long lasting wood construction along with stainless steel handles and hinges provide exceptional resistance to corrosion from harsh acids. Shelf adjusts on 1-1/4" (32-mm) centers to accommodate a wide variety of containers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty