Would you like to log in again?
Whatman™ Quantitative Filter Papers, Ashless Grades (ash 0.01%), Grade 589/1, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10300011 10300045 10300014 10300010 10300009 10300010 10300011 10300012 10300045 10300014 10300012 10300009
14224-460PK 78.14 USD InStock
14224-460 14224-454 CA14224-460 CA11012-880 10034-602 14224-458 CA14224-456 10034-604 CA14224-454 14224-456 CA11012-622 CA14224-458
Whatman™ Quantitative Filter Papers, Ashless Grades (ash 0.01%), Grade 589/1, Whatman products (Cytiva)
Filters Filter Paper Quantitative Filter Paper
With less than 0.01% ash, Cytiva's Whatman Grade 589/1 filter papers can be used for the Blaine test in the cement industry and as an ash content test filter for foodstuffs.
  • Herzberg filtration speed of 50 seconds*
  • Black ribbon filter for fast filtration of coarse precipitates
  • High purity (<0.01% ash)
  • Blaine test and ash content test compatible
  • Also available prepleated as Grade 589/1½
*The time taken to filter 100 mp water (20° C) through a filter area of 10 cm² at a constant pressure of 5 cm water column.
Whatman Grade 589/1 filter papers from Cytiva's business are designed for fast filtration of coarse precipitates in support of tests such as cement fineness measurement in a standard Blaine test or determination of the ash content in foodstuffs.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty