Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Petroleum spirit, 35…60 °C ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Petroleum ether

673838-4X4L
BJ673838-4LCS 1032.77 USD InStock
BJ673838-4L
Petroleum spirit, 35…60 °C ACS, Riedel-de Haën™
Petroleum spirit
  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Boiling Pt: 5.9 %(V)
Melting Pt: -73 °C
Density: 0.64 g/cm³ (20 °C) (lit.)
Flash Pt: -18 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 232-453-7
UN: 1268
ADR: 3,II
Order Now
Specification Test Results
Autoignition Temperature 475 °F
Identity (IR) complying
Refractive index (n 20/D) 1.358 - 1.368
Non-volatile matter Max. 0.001 %
Boiling range 35,0 - 60,0 °C
Vapor Pressure 25.8 psi ( 55 °C)
Acidity complying
APHA Max. 10
Explosion Limit 8 %
Vapor Density 2.5 (vs air)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty