Would you like to log in again?
Support Rings with Clamps, Chemglass
Supplier: Chemglass
CG-3057-10
10034-446EA 41.27 USD InStock
10034-446
Support Rings with Clamps, Chemglass
Supports Flask Supports
Solid ring with built in clamp for attachment directly to support rod.
  • Nickel plated zinc alloy
  • Overall length: 180 mm
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty