Would you like to log in again?
Drip Chamber, ALA Scientific Instruments
Supplier: ALA Scientific Instruments
SPK-DRIP
100133-808EA 112.46 USD InStock
100133-808
Drip Chamber, ALA Scientific Instruments
Filters Filter Tubes
Drip chamber with solution filter (15 μm) and 20 drops/ml for simple perfusion kit.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty