Would you like to log in again?
BTX Multi-Well Electroporation Plates, Harvard Apparatus
Supplier: Harvard Apparatus
BTX
450450-M
10031-730EA 487.64 USD InStock
10031-730
BTX Multi-Well Electroporation Plates, Harvard Apparatus
Electroporation Plates
Run multiple samples in seconds for rapid optimization of electroporation variables and unsurpassed sample through-put.
  • Transfection of Mammalian Cells
  • Transformation of Bacteria and Yeast
  • siRNA and cDNA Library Screening
Complete user control over electroporation settings allows for a wide range of optimization flexibility
Each MOS plate consists of either 96 or 25 individual wells with integrated electrodes. The 96-well plate follows traditional 96-well format i.e., 12 columns of 8 wells each. The 25-well format has 5 columns of 5 wells each. Four different Multi-Well Plates are available: 96-well with 4 mm gap, 96-well with 2 mm gap, 25-well with 4 mm gap and 25-well with 2 mm gap.
Saves valuable lab time, moves experiments further faster, provides reliable results and requires no expensive reagents. Eliminates cross-contamination with patented integrated electrodes and plate sealer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty